Learn More
Bellco Glass, Inc. EZwaste Replacement Fittings, 1/8" MNPT, HB Plug, 25/PK
SDP
Supplier:  Bellco Glass, Inc. 452800169
Encompass_Preferred
The 1/8″ MNPT HB Plug Pack is the replacement fittings for EZwaste Solvent Waste System’s fittings. It includes (25) 1/8″ MNPT Hose Barb Plug. These plugs provide a closed system necessary to prevent spills.The EZwaste Solvent Waste System is designed to be an affordable solution for HPLC liquid waste disposal. Each system is manufactured to safely collect waste from two HPLC machines. This system features an activated carbon filter which traps volatile organic compounds from being released into the air. This prevents chemicals from being inhaled by lab workers and provides an easy mechanism of disposal for vapor wastes. The EZwaste Solvent Waste system also features a closed loop design which protects against leaks and spills, improving overall lab safety. Material: PolyethyleneColor: Black
Catalog No. 50-255-0593