Learn More
Bellco Glass, Inc. Ezwaste UN/DOT HDPE 13.5L Container w/ Closed Cap, Boxed, 1/EA
SDP
Catalog No. 502550503
Encompass_Preferred
13.5L HDPE UN/DOT System is designed to be an affordable solution for liquid waste disposal. Our containers are made to be tough and durable. They are great for secure shipment and storage of liquid products. The UN/DOT container is also available in 13.5L, 20L, and 60L. The 10L UN/DOT containers come with the 50S cap and the 13.5L-60L UN/DOT containers come with the S70 Cap.These containers can also be used in other industries including, but not limited to sanitation, chemical, agricultural and food. All raw materials used in our containers meet FDA Standards for food contact.Made from HDPEMeets UN 3H1 StandardsAvailable in 13.5L, 20L, and 60LUtilizes blow mold technology Height: 11" (280 mm)Length: 11" (280mm)Width: 9" (229 mm)Volume: 13.5LCarboy Material: HDPECap Material: HDPEUN Rating: 3H1/Y1.8/100
Catalog No. 50-255-0503 Supplier Bellco Glass, Inc. Supplier No. 452800259
13.5L HDPE UN/DOT System is designed to be an affordable solution for liquid waste disposal. Our containers are made to be tough and durable. They are great for secure shipment and storage of liquid products. The UN/DOT container is also available in 13.5L, 20L, and 60L. The 10L UN/DOT containers come with the 50S cap and the 13.5L-60L UN/DOT containers come with the S70 Cap.These containers can also be used in other industries including, but not limited to sanitation, chemical, agricultural and food. All raw materials used in our containers meet FDA Standards for food contact.Made from HDPEMeets UN 3H1 StandardsAvailable in 13.5L, 20L, and 60LUtilizes blow mold technology Height: 11" (280 mm)Length: 11" (280mm)Width: 9" (229 mm)Volume: 13.5LCarboy Material: HDPECap Material: HDPEUN Rating: 3H1/Y1.8/100