Learn More
Bellco Glass, Inc. Ezwaste UN/DOT HDPE 20L Container w/ Closed Cap, Boxed, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800260
Encompass_Preferred
20L HDPE UN/DOT System is designed to be an affordable solution for liquid waste disposal. Our containers are made to be tough and durable. They are great for secure shipment and storage of liquid products. The UN/DOT container is also available in 10L, 13.5L, and 60L. The 10L UN/DOT containers come with the 50S cap and the 13.5L-60L UN/DOT containers come with the S70 Cap.These containers can also be used in other industries including, but not limited to sanitation, chemical, agricultural and food. All raw materials used in our containers meet FDA Standards for food contact.Made from HDPEMeets UN 3H1 StandardsAvailable in 13.5L, 20L, and 60LUtilizes blow mold technology Height: 15.5" (394 mm)Length: 11.25" (286mm)Width: 9" (229 mm)Volume: 20LCarboy Material: HDPECap Material: HDPEUN Rating: 3H1/Y1.8/100
Catalog No. 50-255-0502