Learn More
Perkin Elmer US LLC EzyFit Connector for SeaSpray and MicroMist Nebulizers - 0.50 mm I.D. x 1.59 mm O.D. x 700 mm Long Sample Tube, Qty. 10
Catalog No. 502105191
Encompass_Preferred
The EzyFit Twin Sample Tube - 0.50 mm I.D. x 116 in. O.D. x 700 mm Long version (N0777462) of this EzyFit connector can be purchased separately in a pack of 6. This sample tube is replacement tubing for internal standard kits. It is included in the HF-Resistant Internal Standard Kit (N0774067) and the Non HF-Resistant Internal Standard Kit (N0774068).
Catalog No. 50-210-5191 Supplier Perkin Elmer US LLC Supplier No. N0777463
The EzyFit Twin Sample Tube - 0.50 mm I.D. x 116 in. O.D. x 700 mm Long version (N0777462) of this EzyFit connector can be purchased separately in a pack of 6. This sample tube is replacement tubing for internal standard kits. It is included in the HF-Resistant Internal Standard Kit (N0774067) and the Non HF-Resistant Internal Standard Kit (N0774068).