Learn More
Perkin Elmer US LLC EzyFit Connector for SeaSpray and MicroMist Nebulizers - 0.25 mm I.D. x 1.59 mm O.D. x 700 mm Long Sample Tube, Qty. 10
Catalog No. 502105201
Encompass_Preferred
This sample tube is replacement tubing for internal standard kits. It is included in the HF-Resistant Internal Standard Kit (N0774067) and the Non HF-Resistant Internal Standard Kit (N0774068).
Catalog No. 50-210-5201 Supplier Perkin Elmer US LLC Supplier No. N0777464
This sample tube is replacement tubing for internal standard kits. It is included in the HF-Resistant Internal Standard Kit (N0774067) and the Non HF-Resistant Internal Standard Kit (N0774068).