Learn More
Glass Expansion EzyFit Sample Tube 1/16” OD x 0.50mm ID x 300 mm (Pkt 10)
SDP
Catalog No. 502702990
Encompass_Preferred
EzyFit Sample Tube 1/16” OD x 0.50mm ID x 300 mm (Pkt 10). Pack of 10 replacement sample lines to connect Glass Expansion’s Trident CT Internal Standard Kit (P/Ns NC2046960, 60-703-1179) to your Glass Expansion F-Series nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2990 Supplier Glass Expansion Supplier No. EZT1650300
EzyFit Sample Tube 1/16” OD x 0.50mm ID x 300 mm (Pkt 10). Pack of 10 replacement sample lines to connect Glass Expansion’s Trident CT Internal Standard Kit (P/Ns NC2046960, 60-703-1179) to your Glass Expansion F-Series nebulizer.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001