Learn More
Glass Expansion EzyFit with 1.3mm OD x 0.25mm ID x 700mm long sample tube (PKT. 10)
SDP
Catalog No. 502702993
Encompass_Preferred
EzyFit with 1.3mm OD X 0.25mm ID x 700mm long sample tube. Package of 10 replacement sample lines compatible with Glass Expansion F-Series Nebulizers. Used to carry sample from peristaltic pump tubing or mixing T to nebulizer. This consumable part should be replaced at a frequency of about every 6 months to ensure optimum sample flow.For EzyFit Connector with 0.50mm ID, 1/16 OD x 700mm long sample tube, EZT-16-50. For EzyFit Connector with 0.75mm ID, 1.3mm OD x 1500mm long sample tube, EZTS-075.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2993 Supplier Glass Expansion Supplier No. EZT025
EzyFit with 1.3mm OD X 0.25mm ID x 700mm long sample tube. Package of 10 replacement sample lines compatible with Glass Expansion F-Series Nebulizers. Used to carry sample from peristaltic pump tubing or mixing T to nebulizer. This consumable part should be replaced at a frequency of about every 6 months to ensure optimum sample flow.For EzyFit Connector with 0.50mm ID, 1/16 OD x 700mm long sample tube, EZT-16-50. For EzyFit Connector with 0.75mm ID, 1.3mm OD x 1500mm long sample tube, EZTS-075.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001