Learn More
Glass Expansion EzyFit with 1.6mm (1/16”) OD X 0.25mm ID x 700mm long sample tube (PKT 10)
SDP
Catalog No. 502702992
Encompass_Preferred
EzyFit with 1.6mm (1/16”) OD X 0.25mm ID x 700mm long sample tube. Package of 10 replacement sample lines compatible with Glass Expansion F-Series Nebulizers. Used to carry sample from peristaltic pump tubing to nebulizer or to carry internal standard to Glass Expansion’s Trident IS Kit for HF P/N NC2046964, 60-808-1150. This consumable part should be replaced at a frequency of about every 6 months to ensure optimum sample flow.For EzyFit Connector with 0.50mm ID, 1.6mm (1/16”) OD x 700mm long sample tube, EZT-16-50. For EzyFit Connector with 0.75mm ID, 1.3mm OD x 1500mm long sample tube, EZTS-075. For EzyFit Connector with 0.25mm ID, 1.3mm OD x 700mm long sample tube, EZT-025.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-270-2992 Supplier Glass Expansion Supplier No. EZT1625
EzyFit with 1.6mm (1/16”) OD X 0.25mm ID x 700mm long sample tube. Package of 10 replacement sample lines compatible with Glass Expansion F-Series Nebulizers. Used to carry sample from peristaltic pump tubing to nebulizer or to carry internal standard to Glass Expansion’s Trident IS Kit for HF P/N NC2046964, 60-808-1150. This consumable part should be replaced at a frequency of about every 6 months to ensure optimum sample flow.For EzyFit Connector with 0.50mm ID, 1.6mm (1/16”) OD x 700mm long sample tube, EZT-16-50. For EzyFit Connector with 0.75mm ID, 1.3mm OD x 1500mm long sample tube, EZTS-075. For EzyFit Connector with 0.25mm ID, 1.3mm OD x 700mm long sample tube, EZT-025.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001