Learn More
Glass Expansion EzyGlide Lubricated Cloth
SDP
Catalog No. NC1352363
Encompass_Preferred
EzyGlide Lubricated Cloth. Used to lubricate fully demountable torch tubes as it promotes easy insertion/removal. Also useful for lubricating peristaltic pump rollers, where it will reduce pump tubing wear and increases the service life of the tubing.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
This item is not returnable. View return policy
Catalog No. NC1352363 Supplier Glass Expansion Supplier No. 70EZGCLOTH
EzyGlide Lubricated Cloth. Used to lubricate fully demountable torch tubes as it promotes easy insertion/removal. Also useful for lubricating peristaltic pump rollers, where it will reduce pump tubing wear and increases the service life of the tubing.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001