Learn More
Medchemexpress LLC F 13714 fumarate | 208109-39-1 | 99.1% | 10 MG
SDP
Catalog No. 5000340802
Encompass_Preferred
F 13714 fumarate is a selective 5-HT1A receptor biased agonist that exhibits antidepressant-like properties after a single administration in the mouse model of chronic mild stress.
  • Selective 5-HT1A receptor biased agonist.
  • Shows antidepressant-like properties after a single administration in the mouse model of chronic mild stress.
  • Targets 5-HT1A autoreceptors.
  • Normalizes depressive-like behavior in the forced swim test (FST) in UCMS mice.
  • Rescues cortical and hippocampal deficits in p-ERK1/2 levels.
  • Decreases immobility in mice.
  • Decreases rectal body temperature in mice.
  • Possesses antidepressant- and anxiolytic-like properties in naive rats.
Catalog No. 50-003-40802 Supplier Medchemexpress LLC Supplier No. HY12890110MG
F 13714 fumarate is a selective 5-HT1A receptor biased agonist that exhibits antidepressant-like properties after a single administration in the mouse model of chronic mild stress.
  • Selective 5-HT1A receptor biased agonist.
  • Shows antidepressant-like properties after a single administration in the mouse model of chronic mild stress.
  • Targets 5-HT1A autoreceptors.
  • Normalizes depressive-like behavior in the forced swim test (FST) in UCMS mice.
  • Rescues cortical and hippocampal deficits in p-ERK1/2 levels.
  • Decreases immobility in mice.
  • Decreases rectal body temperature in mice.
  • Possesses antidepressant- and anxiolytic-like properties in naive rats.