Learn More
Medchemexpress LLC F 13714 fumarate | 208109-39-1 | MFCD34469533; MFCD11045365 | 99.1% | 538.97 g/mol | C25H29ClF2N4O5 | 5 MG
SDP
Catalog No. 5000262725
Encompass_Preferred
F 13714 fumarate is the fumarate salt of a selective 5-HT1A receptor biased agonist used as a research compound. It has been reported to produce antidepressant-like effects after a single administration in a mouse model of chronic mild stress, and is provided as a well-characterized small molecule for pharmacology and behavioral studies.
  • Selective 5-HT1A receptor biased agonist activity.
  • Demonstrates antidepressant-like effects in preclinical mouse model.
  • Provided as a purified research-grade small molecule.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Characterized by CAS 208109-39-1 and molecular weight 538.97 g/mol.
Catalog No. 50-002-62725 Supplier Medchemexpress LLC Supplier No. HY1289015MG
F 13714 fumarate is the fumarate salt of a selective 5-HT1A receptor biased agonist used as a research compound. It has been reported to produce antidepressant-like effects after a single administration in a mouse model of chronic mild stress, and is provided as a well-characterized small molecule for pharmacology and behavioral studies.
  • Selective 5-HT1A receptor biased agonist activity.
  • Demonstrates antidepressant-like effects in preclinical mouse model.
  • Provided as a purified research-grade small molecule.
  • Suitable for in vitro and in vivo pharmacology studies.
  • Characterized by CAS 208109-39-1 and molecular weight 538.97 g/mol.