Learn More
Medchemexpress LLC F-15599 | 635323-95-4 | 99.1% | 394.85 | 1 ML
SDP
Catalog No. 5000401362
Encompass_Preferred
F-15599 is a highly selective G-protein biased 5-HT1A receptor agonist with a Ki of 3.4 nM, intended for research use. It has been shown to significantly reduce l-DOPA-induced dyskinesia (LID) without affecting motor performance, and to cause a dose-dependent delay in attack latency, reducing aggressive behavior in rats.
  • Acts as a highly selective G-protein biased 5-HT1A receptor agonist.
  • Reduces l-DOPA-induced dyskinesia (LID).
  • Decreases aggressive behavior.
  • Increases discharge rate of pyramidal neurons in the medial prefrontal cortex (mPFC).
  • Modulates dopamine and serotonin release.
Catalog No. 50-004-01362 Supplier Medchemexpress LLC Supplier No. HY1986310MM/1ML
F-15599 is a highly selective G-protein biased 5-HT1A receptor agonist with a Ki of 3.4 nM, intended for research use. It has been shown to significantly reduce l-DOPA-induced dyskinesia (LID) without affecting motor performance, and to cause a dose-dependent delay in attack latency, reducing aggressive behavior in rats.
  • Acts as a highly selective G-protein biased 5-HT1A receptor agonist.
  • Reduces l-DOPA-induced dyskinesia (LID).
  • Decreases aggressive behavior.
  • Increases discharge rate of pyramidal neurons in the medial prefrontal cortex (mPFC).
  • Modulates dopamine and serotonin release.