Learn More
Medchemexpress LLC F-15599 tosylate (NLX-101 tosylate) | 955112-72-8 | 567.05 | 100 MG
SDP
Catalog No. 5000401628
Encompass_Preferred
F-15599 tosylate is a highly selective G-protein biased 5-HT1A receptor agonist, with a Ki of 3.4 nM. It is intended for research use only.
  • Highly selective G-protein biased 5-HT1A receptor agonist
  • Ki of 3.4 nM
  • Related to neurological, eye or ear disease research
  • Can be found in compound screening libraries
Catalog No. 50-004-01628 Supplier Medchemexpress LLC Supplier No. HY19863100MG
F-15599 tosylate is a highly selective G-protein biased 5-HT1A receptor agonist, with a Ki of 3.4 nM. It is intended for research use only.
  • Highly selective G-protein biased 5-HT1A receptor agonist
  • Ki of 3.4 nM
  • Related to neurological, eye or ear disease research
  • Can be found in compound screening libraries