Learn More
MSC F/24 30 FAN HEAD PULL SWITCH
Catalog No. 5000505964
Encompass
This fan speed control is designed to regulate the operational speed of multiple fans, providing adjustable control for optimal performance. It supports a range of 6 to 10 fans.
  • Controls speed of 6 to 10 fans
  • Operates at 115 V
  • Amperage rating of 1.5 A
Catalog No. 50-005-05964 Supplier MSC Supplier No. 38857835
This fan speed control is designed to regulate the operational speed of multiple fans, providing adjustable control for optimal performance. It supports a range of 6 to 10 fans.
  • Controls speed of 6 to 10 fans
  • Operates at 115 V
  • Amperage rating of 1.5 A