Learn More
MSC Mag-Mate Grinder Holder, 3-6 in Angle Grinder Compatible, Stainless Steel, Permanent Ceramic Magnet
Catalog No. 5000536352
Encompass
This grinder holder is designed to keep angle grinders handy on workbenches. It features a sturdy stainless steel construction and a powerful permanent ceramic magnet base, allowing for quick placement and repositioning. It is compatible with 3-6 in electric and air-powered angle grinders, holding them in horizontal or vertical positions.
  • Sturdy stainless steel holder
  • Plated permanent ceramic magnet
  • Magnetic base allows for quick placement and repositioning
  • Holds 3-6 in angle grinders
  • Compatible with electric and air powered grinders
  • Two attachment options available with screws or magnets
  • Holds grinder wheels in horizontal or vertical positions
Catalog No. 50-005-36352 Supplier MSC Supplier No. 16300584
This grinder holder is designed to keep angle grinders handy on workbenches. It features a sturdy stainless steel construction and a powerful permanent ceramic magnet base, allowing for quick placement and repositioning. It is compatible with 3-6 in electric and air-powered angle grinders, holding them in horizontal or vertical positions.
  • Sturdy stainless steel holder
  • Plated permanent ceramic magnet
  • Magnetic base allows for quick placement and repositioning
  • Holds 3-6 in angle grinders
  • Compatible with electric and air powered grinders
  • Two attachment options available with screws or magnets
  • Holds grinder wheels in horizontal or vertical positions