Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ F(ab')2-Donkey anti-Goat IgG (H+L) Cross-Adsorbed Secondary Antibody, PE

Catalog No. PI31860 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI31860 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI31860 Supplier Invitrogen™ Supplier No. 31860
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

F(ab')2-Donkey Polyclonal Secondary Antibody

Concentration may vary slightly from lot-to-lot, see lot-specific datasheet for exact concentration. Product # 31860 has been successfully used in Western blot, IF, ICC, IHC, IP and FACS applications. Product # 31860 reacts with the heavy chains of goat IgG and with light chains common to most goat immunoglobulins, but does not react against non-immunoglobulin serum proteins. This antibody has been tested by ELISA and/or solid-phase adsorbed to ensure minimal cross-reaction with chicken, guinea pig, Syrian hamster, horse, human, mouse, rabbit and rat serum proteins. However, these antibodies may cross-react with immunoglobulins from other species. Phycobiliprotein Concentration: 0.5 mg/mL (determined by absorption = 82.0 at 566 nm for a 1% solution for only those R-PE molecules to which at least one molecule of active antibody is bound) Phycoerythrin Source: Seaweed Store product protected from light at 4°C until opened. DO NOT FREEZE. RPE Amax= 490 nm, 545 nm, and 566 nm; Emax= 580 nm. Reconstitute with 1.0 mL of distilled water (0.5 mg/mL after restoration). Country of Origin: USA.

Anti-Goat secondary antibodies are affinity-purified antibodies with well-characterized specificity for goat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Goat IgG (H+L) Cross-Adsorbed
Applications Flow Cytometry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate PE
Formulation PBS with 15mg/mL BSA and 0.05% sodium azide; pH 7.6
Host Species Donkey
Purification Method Antigen affinity chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Goat
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.