Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

F(ab')2 Fragment Rabbit anti-Swine IgG (Whole Molecule), HRP, Secondary Antibody, MP Biomedicals™

Catalog No. MP0855826 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP0855826 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP0855826 Supplier MP Biomedicals, Inc Supplier No. 0855826/DEL

missing translation for 'validMainframeMsg'

RabbitF(ab')2 Fragment to Swine IgG (Whole Molecule) Secondary Antibody

  • Product is the lyophilized powder of horseradish peroxidase (HRP)-conjugated rabbit F(ab')2 fragment to swine IgG (whole molecule) and buffer salts.
  • This product is suitable for use as a reagent in enzyme immunoassays (EIA), cell and tissue staining (for light microscopy), cell and tissue labeling (for electron microscopy), and blot immunostaining.
TRUSTED_SUSTAINABILITY

Specifications

Applications Immunohistochemistry
Classification Polyclonal
Concentration Total Protein: 19.0mg/mL; Titer: 1:88,000
Conjugate HRP
Description Product is the lyophilized powder of horseradish peroxidase (HRP)-conjugated rabbit F(ab')2 fragment to swine IgG (wholemolecule) and buffer salts.
Formulation 0.02M sodium phosphate, 0.14M sodium chloride, pH 7.3,
Host Species Rabbit
Immunogen Purified Pig IgG
Purification Method salt fractionation
Quantity 2 mL
Regulatory Status RUO
Primary or Secondary Secondary
Reconstitution Reconstitute product with 2.0mL of deionized or distilled water.gentle swirling may be used to speed rehydration. Avoid vigorous shaking of the reconstituted material.
Target Species Swine
Content And Storage Store lyophilized product at 2-8°C. For long-term storage, aliquot and freeze reconstituted product at -20°C or lower.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.