Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ F(ab')2-Goat anti-Human IgG (H+L) Cross-Adsorbed Secondary Antibody, HRP

Catalog No. PIPA533290 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
600 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533290 600 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533290 Supplier Invitrogen™ Supplier No. PA533290
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

F(ab')2-Goat Polyclonal Secondary Antibody

PA5-33290 targets Human IgG in ELISA and IHC (F) applications. This antibody does not cross-react with Goat IgG F(c) or mouse, rat, bovine and horse serum proteins, as determined by immunoelectrophoresis. F(ab')2 fragments were prepared by pepsin digestion.

Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Human IgG (H+L) Cross-Adsorbed
Applications ELISA, Immunohistochemistry (Frozen)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate HRP
Formulation PBS with 0.01% thimerosal
Host Species Goat
Immunogen Human IgG.
Purification Method Purified
Quantity 600 μg
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.