Learn More
Invitrogen™ F(ab')2-Goat anti-Human Kappa Light Chain Secondary Antibody, PE
Shop All Thermo Scientific ProductsDescription
This antibody has been cross-absorbed with pooled human myeloma proteins with lambda light chains. Purified IgG was prepared from whole serum by affinity chromatography, and F(ab)2 fragments were prepared by pepsin digestion. For FACS analysis, use 10 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Goat F(ab')2 anti Human Kappa Light Chain antibody recognizes human kappa light chains. This antibody has been cross absorbed with pooled human myeloma proteins with lambda light chains.
Specifications
Specifications
| Antigen | Human Kappa Light Chain |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Concentration | 0.25 mg/mL |
| Conjugate | PE |
| Formulation | PBS with proprietary stabilizer and <0.1% sodium azide; pH 7.4 |
| Gene Alias | Iglc |
| Host Species | Goat |
| Immunogen | Human kappa light chains. |
| Purification Method | Affinity chromatography |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.