Learn More
Invitrogen™ F(ab')2-Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody
Description
F(ab')2-Goat anti-Mouse IgG (H+L) Highly Cross-Adsorbed Secondary Antibody for ELISA
Specifications
Specifications
| Antigen | Mouse IgG (H+L) Highly Cross-Adsorbed |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | >4.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.05% sodium azide; pH 7.2 |
| Host Species | Goat |
| Immunogen | Gamma Immunoglobins Heavy and Light chains. |
| Purification Method | Affinity Chromatography |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.