Learn More
F(ab')2-Goat anti-Mouse IgG (H+L) Secondary Antibody, Qdot™ 605, Invitrogen™
Catalog No. 502544944
Encompass_Preferred
Catalog No. 50-254-4944 Supplier Invitrogen Supplier No. 502544944
Only null left
Add to Cart
Add to Cart
F(ab')2-Goat Polyclonal Secondary Antibody
Qdot nanocrystals are composed of semi-conductor material to generate a fluorescent particle which is exceptionally bright and does not photobleach. Qdot nanocrystals paired with the correct optical filters are as much as 50 times brighter than traditional organic dyes.
Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.