Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ F(ab')2-Goat anti-Rabbit IgG (H+L) Secondary Antibody, NANOGOLD™

Catalog No. N24916
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N24916 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N24916 Supplier Invitrogen™ Supplier No. N24916
Add to Cart
Add to Cart

F(ab')2-Goat Polyclonal Secondary Antibody

The NANOGOLD conjugate of Fab' fragment of goat anti-rabbit IgG is labeled with small uniformly sized (1.4 nm) gold particles at a ~1:1 ratio in the hinge region, remote from the antigen-binding site and can be used in immunoblotting, light microscopy, and electron microscopy applications.

Anti-Rabbit secondary antibodies are affinity-purified antibodies with well-characterized specificity for rabbit immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit IgG (H+L)
Applications Immunomicroscopy, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Nanogold
Formulation PBS with 0.1% BSA and 0.05% sodium azide; pH 7.4
Host Species Goat
Immunogen Gamma Immunoglobin.
Purification Method Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage 4°C
Product Type Secondary Antibody
Form Liquid
Isotype IgG
Show More Show Less
I want to label F-actin with phalloidin and image with transmission electron microscopy. What is the best way to go about it?

One way is to use a biotin-phalloidin conjugate, then label that with a colloidal gold conjugate of streptavidin. You will need to block endogenous biotin first. Another method is to label with fluorescein phalloidin, then with an anti-fluorescein antibody conjugated with colloidal gold.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.