Learn More
Invitrogen™ F(ab')2-Goat anti-Rat IgG (H+L) Cross-Adsorbed Secondary Antibody, PE
Shop All Thermo Scientific ProductsDescription
F(ab')2 fragments were prepared by pepsin digestion. Cross-reactivity with mouse IgG has been reduced by adsorption. For FACS analysis, use 50 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Reconstitute with 0.5 mL of distilled water. After reconstitution, store product undiluted at 4°C in the dark.
Specifications
Specifications
| Antigen | Rat IgG (H+L) Cross-Adsorbed |
| Applications | Flow Cytometry |
| Classification | Polyclonal |
| Concentration | Conc. Not Determined |
| Conjugate | PE |
| Formulation | PBS with 5% sucrose, 1% BSA and 0.09% sodium azide |
| Host Species | Goat |
| Immunogen | whole rat IgG. |
| Purification Method | Purified |
| Quantity | 500 μL |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.