Learn More
Rockland Immunochemicals F(ab')2 Goat IgG (H&L) Antibody Phycoerythrin Conjugated Pre-Adsorbed 500 µg
SDP
Supplier:  Rockland Immunochemicals 705708125
Encompass
F(ab')2 Anti-Goat IgG Phycoerythrin Antibody was generated by enzymatic cleavage and subsequent separation from the Fc fragment. Because of their smaller size F(ab)2 fragments offer several advantages over intact antibodies for use in certain immunochemical techniques and experimental applications. F(ab)2 fragments penetrate tissue samples and show better antigen recognition and signal generation in IHC. F(ab)2 fragments lack the Fc region and therefore do not bind Fc receptors which effectively lowers background staining. F(ab')2 Antibody is ideal for investigators who routinely perform flow cytometry immunohistochemistry or IHC and other immunoassays.
Catalog No. 50-194-3943