Learn More
Rockland Immunochemicals F(ab')2 Human κ (kappa chain) Antibody Fluorescein Conjugated 1 mg
SDP
Supplier:  Rockland Immunochemicals 7091210
Encompass
F(ab')2 HUMAN kappa Antibody Fluorescein Conjugated is designed for immunofluorescence microscopy fluorescence based plate assays (FLISA) and fluorescent western blotting. This product is also suitable for multiplex analysis including multicolor imaging utilizing various commercial platforms.
Catalog No. 50-194-3946