Learn More
Rockland Immunochemicals F(ab')2 Human IgM Fc5µ Antibody Biotin Conjugated 500 µg
SDP
Supplier:  Rockland Immunochemicals 7091631
Encompass
F(ab')2 Anti-Human IgM Fc5µ Biotin Antibody generated in goat detects specifically the Fc5μ portion of the human IgM heavy chain. Immunoglobulin M is the largest antibody isotype and the first to be secreted against an initial exposure to antigen. IgM is predominantly produced in the spleen. Formed from covalently linking 5 immunoglobulins together. F(ab')2 Anti-Human IgM Fc5µ antibody is ideal for investigators in Immunology Microbiology and Cell Biology.
Catalog No. 50-194-3947