Learn More
Rockland Immunochemicals F(ab')2 Mouse IgG F(c) Antibody Peroxidase Conjugated Pre-Adsorbed 500 µg
SDP
Supplier:  Rockland Immunochemicals 7101322
Encompass
F(ab')2 Anti-Mouse IgG F(c) Peroxidase Antibody was generated in goat and detects specifically Mouse IgG F(c). Secondary Antibodies are available in a variety of formats and conjugate types. When choosing a secondary antibody product consideration must be given to species and immunoglobulin specificity conjugate type fragment and chain specificity level of cross-reactivity and host-species source and fragment composition. F(ab')2 Anti-Mouse IgG F(c) Antibody is ideal for investigators in Cancer Immunology and Microbiology research.
Catalog No. 50-194-3950