Learn More
Invitrogen™ F(ab')2-Rabbit anti-Mouse IgG (H+L) Cross-Adsorbed Secondary Antibody, FITC
Shop All Thermo Scientific ProductsDescription
Some cross reactivity with mouse IgM and IgA is expected, as is cross reactivity with rat IgG. Cross reactivity with human serum proteins has been minimized by solid phase adsorption. F(ab')2 fragments were prepared by pepsin digestion of the IgG followed by a gel filtration step to remove the remaining intact IgG or Fc fragments. For Flow Cytometry, use 50 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. FITC conjugated Rabbit F(ab')2 anti Mouse IgG antibody recognizes all subclasses of mouse IgG.
Specifications
Specifications
| Antigen | Mouse IgG (H+L) Cross-Adsorbed |
| Applications | Flow Cytometry, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with 0.09% sodium azide; pH 7.2 |
| Host Species | Rabbit |
| Immunogen | Purified murine IgG. |
| Purification Method | Affinity chromatography |
| Quantity | 1 mg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.