Learn More
Invitrogen™ F(ab')2-Rabbit anti-Rat IgG (H+L) Secondary Antibody, HRP
Shop All Thermo Scientific ProductsDescription
PA1-29927 detects Rat IgG (H+L) from rat samples.
Specifications
Specifications
| Antigen | Rat IgG (H+L) |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | HRP |
| Formulation | PBS with 1% BSA and 0.01% gentamicin sulfate; pH 7.2 |
| Host Species | Rabbit |
| Immunogen | Rat IgG whole molecule. |
| Purification Method | IgG fraction |
| Quantity | 250 μg |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.