Learn More
Rockland Immunochemicals F(ab')2 Rabbit IgG [H&L] Antibody Phycoerythrin conjugated Pre-adsorbed 500 µg
SDP
Supplier:  Rockland Immunochemicals 711708127
Encompass
F(ab')2 Anti-Rabbit IgG [H&L] Antibody Phycoerythrin Conjugated Pre-adsorbed was generated by enzymatic cleavage and subsequent separation from the Fc fragment. Because of their smaller size F(ab)2 fragments offer several advantages over intact antibodies for use in certain immunochemical techniques and experimental applications. F(ab)2 fragments penetrate into tissue samples and show better antigen recognition and signal generation in IHC. F(ab)2 fragments lack the Fc region and therefore do not bind Fc receptors which effectively lowers background staining. F(ab')2 Rabbit IgG [H&L] Antibody Phycoerythrin conjugated Pre-adsorbed is ideal for investigators who routinely perform flow cytometry immunohistochemistry or IHC and other immunoassays.
Catalog No. 50-194-3953