Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ F(ab)-Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, Alexa Fluor™ Plus 488

Catalog No. 5000804155
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-008-04155 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-008-04155 Supplier Invitrogen™ Supplier No. SA510397AFP488
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

F(ab)-Goat Recombinant Superclonal Secondary Antibody

F(ab)-Goat anti-Mouse IgG (H+L) Superclonal™ Recombinant Secondary Antibody, Alexa Fluor™ Plus 488 can detect all subtypes of Mouse IgG (Mouse IgG1,2a,2b). It showed minimum to no cross-reactivity to various species immunoglobulins tested. This Fab fragment antibody is expressed in vitro and purified as a recombinant construct. It is also engineered to deliver enhanced stability and sensitivity. The advantages of recombinant antibodies include better specificity, animal origin-free formulation, and lot-to-lot consistency. F(ab) fragments offer several advantages over full-length IgGs due to their smaller molecular size (~50 kDa vs. ~150 kDa), such as improved tissue penetration. The absence of the Fc region reduces nonspecific binding and background staining in Fc receptor-expressing cells and tissues, while also enabling increased flexibility in multiplexing assays. Superclonal™ recombinant secondary antibodies are produced using a multifaceted clonal selection process that involves several phenotypic screens to ensure enhanced specificity, animal origin free formulation and lot-to-lot consistency. Alexa Fluor™ Plus dyes have been chemically optimized to provide higher signal to noise ratios and minimized cross-reactivity to help detect low-abundance targets in rare or precious samples. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

Anti-Mouse secondary antibodies are affinity-purified antibodies with well-characterized specificity for mouse immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Mouse IgG (H+L)
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Superclonal
Concentration 1 mg/mL
Conjugate Alexa Fluor Plus 488
Formulation Proprietary buffer with 0.008% Bromonitrodioxane, 0.008% Methylisothiazolone; pH 6.3-7.3
Host Species Goat
Immunogen Purified Mouse IgG.
Purification Method Affinity chromatography
Quantity 250 μL
Regulatory Status RUO
Primary or Secondary Secondary
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Secondary Antibody
Form Liquid
Isotype IgG F(ab)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.