Learn More
Colonial Medical Supply Co Inc F/AIR Canister, Absorbs Halogenated Anesthetic Gases, Non-Hazardous Design, 8 Canisters/Case
SDP
Catalog No. NC2034480
Encompass
The F/AIR Canister is a disposable anesthesia gas filter solution designed to efficiently eliminate waste anesthetic gases (WAGs) in medical and veterinary settings. Using activated carbon, it effectively absorbs halogenated anesthetic agents, thus enhancing safety by releasing purified air. Its compact design allows for easy storage, and the canister's passive scavenging system simplifies post-procedure cleanup.
Catalog No. NC2034480 Supplier Colonial Medical Supply Co Inc Supplier No. B80120AM
The F/AIR Canister is a disposable anesthesia gas filter solution designed to efficiently eliminate waste anesthetic gases (WAGs) in medical and veterinary settings. Using activated carbon, it effectively absorbs halogenated anesthetic agents, thus enhancing safety by releasing purified air. Its compact design allows for easy storage, and the canister's passive scavenging system simplifies post-procedure cleanup.