Learn More
Bellco Glass, Inc. Flast Bottom Canister Complete, 38X130mm
SDP
Catalog No. 501944670
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom pad prevents chipping and breaking of pipette tips. All components can and will withstand repeated dry or steam heat serialization. For Pasteur pipettes - 5 3/4" long. MADE IN THE USA Case qty-6
Catalog No. 50-194-4670 Supplier Bellco Glass, Inc. Supplier No. 129038130
Manufactured from type 1 class a Borosilicate glass. Bottom pad prevents chipping and breaking of pipette tips. All components can and will withstand repeated dry or steam heat serialization. For Pasteur pipettes - 5 3/4" long. MADE IN THE USA Case qty-6