Learn More
Bellco Glass, Inc. Flat Bottom Canister cComplete, 38X250mm
SDP
Catalog No. 501944671
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Bottom pad prevents chipping and breaking of pipette tips. All components can and will withstand repeated dry or steam heat serialization. Supplied stainless steel closure. MADE IN THE USA Case qty-6
Catalog No. 50-194-4671 Supplier Bellco Glass, Inc. Supplier No. 129038250
Manufactured from type 1 class a Borosilicate glass. Bottom pad prevents chipping and breaking of pipette tips. All components can and will withstand repeated dry or steam heat serialization. Supplied stainless steel closure. MADE IN THE USA Case qty-6