Learn More
Bellco Glass, Inc. Flat Bottom Canister Only, 38X250mm
SDP
Catalog No. 501944667
Encompass_Preferred
FLAT BOTTOM GLASS PIPETTE CANISTER W/SILICONE PAD, MANUFACTURED FROM BOROSILICATE GLASS. ALL COMPONENTS WILL WITHSTAND REPEATED DRY OR STEAM HEAT STERILIZATION BOTTOM PAD PREVENTS CHIPPING OF PIPETTE TIPS. MADE IN THE USA CASE Qty: 6
Catalog No. 50-194-4667 Supplier Bellco Glass, Inc. Supplier No. 129000250
FLAT BOTTOM GLASS PIPETTE CANISTER W/SILICONE PAD, MANUFACTURED FROM BOROSILICATE GLASS. ALL COMPONENTS WILL WITHSTAND REPEATED DRY OR STEAM HEAT STERILIZATION BOTTOM PAD PREVENTS CHIPPING OF PIPETTE TIPS. MADE IN THE USA CASE Qty: 6