Learn More
Bellco Glass, Inc. Flat Bottom Flange Vessel, 3L Bioreactor
SDP
Catalog No. 501944876
Encompass_Preferred
Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. MADE IN THE USA. 3L.
Catalog No. 50-194-4876 Supplier Bellco Glass, Inc. Supplier No. 196488030
Heavy walled construction for cell culture and fermentation applications. Heavy duty o-ring flange creates positive seal for stainless steel head-plate. For use with stainless steel head-plate, o-ring, and locking clamp. MADE IN THE USA. 3L.