Learn More
Bellco Glass, Inc. Flat Bottom Vessel,28X94mm
SDP
Catalog No. 501945780
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Flat bottom borosilicate glass tube with 32mm thread opening. Screened with a 10mm white marking spot. Port assembly is manufactured from 316L stainless steel. Electropolished for easy cleaning. The Long tube extends approximately 25mm above/below the cap. The short tube extends 25mm above the cap. MADE IN THE USA
Catalog No. 50-194-5780 Supplier Bellco Glass, Inc. Supplier No. 563728094
Manufactured from Type 1 Class A Borosilicate Glass. Flat bottom borosilicate glass tube with 32mm thread opening. Screened with a 10mm white marking spot. Port assembly is manufactured from 316L stainless steel. Electropolished for easy cleaning. The Long tube extends approximately 25mm above/below the cap. The short tube extends 25mm above the cap. MADE IN THE USA