Learn More
Bellco Glass, Inc. Flat Bottom Vessel Complete, 32mm
SDP
Catalog No. 501945791
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Flat bottom Borosilicate glass tube with 32mm thread opening. Screened with a 10mm white marking spot. -TUBE MEASURES APPROXIMATELY 32MM OD X 94MM OAL Port assembly is manufactured from 316L stainless steel. Electropolished for easy cleaning. -LONG TUBE EXTENDS APPROXIMATELY 25MM ABOVE/BELOW THE CAP;SHORT TUBE EXTENDS 25MM ABOVE THE CAP. -SUPPLIED WITH A 32MM OPEN TOPBLUE POLYPROPYLENE CAP & WHITE SILICONE SEALING GASKET. MADE IN THE USA
Catalog No. 50-194-5791 Supplier Bellco Glass, Inc. Supplier No. 563752894
Manufactured from Type 1 Class A Borosilicate Glass. Flat bottom Borosilicate glass tube with 32mm thread opening. Screened with a 10mm white marking spot. -TUBE MEASURES APPROXIMATELY 32MM OD X 94MM OAL Port assembly is manufactured from 316L stainless steel. Electropolished for easy cleaning. -LONG TUBE EXTENDS APPROXIMATELY 25MM ABOVE/BELOW THE CAP;SHORT TUBE EXTENDS 25MM ABOVE THE CAP. -SUPPLIED WITH A 32MM OPEN TOPBLUE POLYPROPYLENE CAP & WHITE SILICONE SEALING GASKET. MADE IN THE USA