Learn More
Bio IVT F BGLDOG CRY HEP 5 M CLLSV
Catalog No. 502310769
Encompass
Hepatocyte, Cryopreserved; Celsis In Vitro Technologies; From female Beagle dog, pooled; Storage temperature:-150 deg.C; Used to study phase I and phase II metabolism; Size: 5 million cells per vial
Catalog No. 50-231-0769 Supplier Bio IVT Supplier No. F00205
Hepatocyte, Cryopreserved; Celsis In Vitro Technologies; From female Beagle dog, pooled; Storage temperature:-150 deg.C; Used to study phase I and phase II metabolism; Size: 5 million cells per vial