Learn More
Kuhner Shaker Inc F-SIZED DUAL TRAY WITH 200MM S
Supplier:  Kuhner Shaker Inc SM106416
Encompass
F-Sized dual tray with 200mm spacing between top and bottom levels. Includes polycarbonate plate and sticky strips on each level. For use with flasks up to 250ml in Kuhner Shaker platforms only. Shake speed and application limitations apply - please contact usoffice@kuhner.com for additional details.
Catalog No. 50-257-0318
Add to Cart
Request a Quote