Learn More
Kuhner Shaker Inc F-SIZED DUAL TRAY WITH 200MM S
Catalog No. 502570312
Encompass
F-Sized dual tray with 200mm spacing between top and bottom levels. Includes polycarbonate plate and large sticky mats on each level. For use with flasks up to 250ml in Kuhner Shaker platforms only. Shake speed and application limitations apply - please contact usoffice@kuhner.com for additional details.
Catalog No. 50-257-0312 Supplier Kuhner Shaker Inc Supplier No. SM107413
Add to Cart
Add to Cart
Request a Quote
F-Sized dual tray with 200mm spacing between top and bottom levels. Includes polycarbonate plate and large sticky mats on each level. For use with flasks up to 250ml in Kuhner Shaker platforms only. Shake speed and application limitations apply - please contact usoffice@kuhner.com for additional details.