Learn More
Thermo Scientific™ F1-ClipTip™ Variable Volume Single Channel Pipettes
GSA_VAGREENER_CHOICE
Catalog No. 14488016N
Click to view available options
Volume (Metric):
0.1 to 2 μL
1 to 10 μL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
5 to 50 μL
Compatible Tips:
ClipTip 1000
ClipTip 12.5 Ext
ClipTip 20
ClipTip 200
ClipTip 300
ClipTip 50
Lock tips firmly to the pipette with a complete seal with ClipTip technology, so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces.
Ensure a secure fit with ClipTip technology, which locks tips firmly to the pipette with a complete seal so they will not loosen or fall off. Lightweight handle design combined with minimal tip attachment and ejection forces will increase the conform in pipetting. Feel the difference.
Worry-free pipetting is finally here with the only tip that clips.
Sealed in security of ClipTip Technology. Feel the difference.
Eliminate the frustration of banging tips on your pipette
With a light touch, each tip is clipped securely on the pipette.
Reduce the risk of loose or leaking tips
When ClipTip pipette tips are on, they are sealed and will not loosen. The ClipTip pipetting system remains sealed until you decide to eject tips, even in the most comprehensive applications, including multiple tip touches and mixing cycles.
Enable consistent and reproducible pipetting from user to user
Higher-quality results and more efficient research. Learn more
Ergonomically In-touch
Lightweight handle design combined with low plunger, tip attachment and ejection forces help reduce the risk of repetitive strain injuries
Secure Volume Lock
The secure volume setting prevents undesirable volume drift during operation. Simply lift the plunger up to set volume and press down to lock in place with minimum effort.
Adjustable Comfort
The 120° adjustable finger rest – for both right- and left-handed operator comfort – allows the hand to relax during pipetting cycles.
Improved small volume pipetting
New 0.1-2 μL model and improved versions 1-10 μL models allow optimized performance in your small volume applications.
Wide Selection of ClipTip Pipette Tips
Compatible sizes with F1-ClipTip pipettes: 12.5 Ext, 20, 50, 200, 300, 300 Ext and 1000μL
Life-time Warranty
With convenient online registration, and may vary by country.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips ClipTip 200
Description F1-ClipTip
Color Yellow
Number of Channels 1
Product Line F1-ClipTip
Type Variable Volume Pipette
Volume (Metric) 10 to 100 μL
Increments 0.2μL
Color Code Yellow
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.