Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ F1-ClipTip™ Variable Volume Single Channel Pipettes: PROMO

Catalog No. p-7078194 Shop All Thermo Scientific Products
Change view
Click to view available options
Volume (Metric):
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL
30 to 300 μL
5 to 50 μL
Color:
Blue
Orange
Pink
Violet
Yellow
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color
14-488-014TI 2 to 20 μL Pink
14-488-015TI 5 to 50 μL Violet
14-488-016TI 10 to 100 μL Yellow
14-488-017TI 20 to 200 μL Yellow
14-488-018TI 30 to 300 μL Orange
14-488-019TI 100 to 1000 μL Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 14-488-014TI Supplier Thermo Scientific™ Supplier No. P4641180
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Streamline daily pipetting with innovative Thermo Scientific™ ClipTip™ interlocking technology, which ensures a complete seal with minimal tip attachment.

Innovative ClipTip interlocking tip interface technology
  • Tip is locked and sealed in place until ejected— no dropped tips that waste valuable samples
  • Complete seal to secure the sample volume in each tip for enhanced accuracy and precision
  • Tips are attached and ejected with a light touch— eliminates the frustration of banging tips on the pipette

  • Ergonomic design

  • Offering low attachment, ejection and plunger forces for safe and comfortable use
  • Remarkably lightweight

  • Modern design protects user from the risk of Repetitive Strain Injuries (RSI)
  • Adjustable comfort

  • The 120° adjustable finger rest—for both right- and left-handed operator comfort—allows the hand to relax during pipetting cycles
  • Super blow-out

  • 150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action†
  • Soft-touch tip ejection

  • Innovative gearing mechanism produces more downward force with less effort, releasing tips with a gentle push
  • Volume Lock Adjustment

  • The secure volume setting prevents undesirable volume drift during operation
  • Industry-leading lifetime warranty

  • With convenient online registration
  • Ordering Information:

    † Available in 50μL models and below

    Compatible with:

    Thermo Scientific ClipTip pipette tips.

    TRUSTED_SUSTAINABILITY

    Specifications

    Accuracy 1 to 3%
    Certifications/Compliance The F1-ClipTip pipetting system is CE-marked in accordance with IVD Directive in Europe. Manufactured according to ISO 9001, ISO 14001 and ISO 13485.
    ClipTip 20
    Compatible Tips F1-ClipTip 20
    Color Pink
    Model F1-ClipTip™
    Number of Channels 1
    Product Line F1-ClipTip
    Type Variable Volume Pipette
    Volume (Metric) 2 to 20 μL
    Increments 0.02 μL
    Precision 0.4 to 2.5%
    Show More Show Less

    Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.