Learn More
Mettler Toledo™ F1 OIML Class Standard Weight Set (F1 AL C)
Catalog No. 01912624 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Weight: 1 mg to 200 g
Catalog No.
01-912-624 Weight: 1 mg to 200 g
1 options
Catalog No. 01-912-624 Supplier Mettler Toledo™ Supplier No. 158907
Add to Cart
Add to Cart
F1 OIML class weight set housed in an aluminum box includes accessories for handling and cleaning, and a calibration certificate.
  • With calibration certificate
  • Traceability Certificate: Delivered with a Calibration Certificate in accordance with ISO/IEC 17025 requirements
  • Weight Calibration Performed by Robots: State-of-the-art robot technology minimizes errors and increases accuracy
  • High-grade stainless steel: Low magnetic permeability and high corrosion resistance ensure reliable values
TRUSTED_SUSTAINABILITY
Specifications
Product Type Test Weight
Material Stainless Steel
Case Material Plastic
Weight (Metric) 1 mg to 200 g
Density 8000 (± 60) kg/m3
Includes 23 Pieces; With certificate