Learn More
Aviva Systems Biology Corp F10 Antibody - C-terminal region (ARP41753_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41753_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against F10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human F10. Alternate names: FX, FXAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-57982