Learn More
Alkali Scientific F1001 Smart-Q 100 Fluorometer
SDP
Catalog No. 503029587
Encompass_Preferred
Smart-Q Fluorometer for quantification of DNA RNA and protein samplesSensitive quantitation down to 0 1pg/ul ds DNALarge color touch screen for programming and operationTwo models for single or 8-sample processingBuilt-in reagent calculators for sample preparationCompact and portable (11 x 6 inch footprint)The Alkali ScientificSmart-Q100 Smart-Q800 Fluorometers are designed for the precise quantification of DNA RNA and protein samples By utilizing well-established methodologies for fluorescence detection the Smart-Q Fluorometers provide higher specificity and enhanced accuracy compared to the traditional method of UV-absorbance detection Accurate results are particularly crucial in workflows involvingNext Generation SequencingPCRTransfectionWestern blottingImmunoassaysThe Smart-Q Fluorometers are engineered to achieve optimal performance with Alkali Scientific Fluorescent DNA Quantification Kits and assay tubes however they also maintain compatibility with a range of k
Catalog No. 50-302-9587 Supplier Alkali Scientific Supplier No. SQ1001
Smart-Q Fluorometer for quantification of DNA RNA and protein samplesSensitive quantitation down to 0 1pg/ul ds DNALarge color touch screen for programming and operationTwo models for single or 8-sample processingBuilt-in reagent calculators for sample preparationCompact and portable (11 x 6 inch footprint)The Alkali ScientificSmart-Q100 Smart-Q800 Fluorometers are designed for the precise quantification of DNA RNA and protein samples By utilizing well-established methodologies for fluorescence detection the Smart-Q Fluorometers provide higher specificity and enhanced accuracy compared to the traditional method of UV-absorbance detection Accurate results are particularly crucial in workflows involvingNext Generation SequencingPCRTransfectionWestern blottingImmunoassaysThe Smart-Q Fluorometers are engineered to achieve optimal performance with Alkali Scientific Fluorescent DNA Quantification Kits and assay tubes however they also maintain compatibility with a range of k