Learn More
Medchemexpress LLC F11R peptide TFA | 97.6% | 1459.58 | 5 MG
SDP
Catalog No. 5000422242
Encompass_Preferred
F11R peptide TFA is an F11 receptor molecule that inhibits anti-F11R antibody-induced platelet aggregation and the adhesion of platelets to cytokine (TNFα and INF-γ)-inflamed endothelial cells. It can be used for research of atherosclerotic plaques and associated thrombotic disease.
  • Inhibits anti-F11R antibody-induced platelet aggregation.
  • Inhibits the adhesion of platelets to cytokine (TNFα and INF-γ)-inflamed endothelial cells.
Catalog No. 50-004-22242 Supplier Medchemexpress LLC Supplier No. HYP109185MG
F11R peptide TFA is an F11 receptor molecule that inhibits anti-F11R antibody-induced platelet aggregation and the adhesion of platelets to cytokine (TNFα and INF-γ)-inflamed endothelial cells. It can be used for research of atherosclerotic plaques and associated thrombotic disease.
  • Inhibits anti-F11R antibody-induced platelet aggregation.
  • Inhibits the adhesion of platelets to cytokine (TNFα and INF-γ)-inflamed endothelial cells.