Learn More
ATCC F-12K Medium
Catalog No. 502383973
Encompass
F-12K Medium (Kaighn's Modification of Ham's F-12 Medium) contains 2 mM L-glutamine and 1500 mg/L sodium bicarbonate. It has applications for cell culture, cell growth, and viability.
Catalog No. 50-238-3973 Supplier ATCC Supplier No. 302004
 Restricted Item
Add to Cart
Add to Cart
F-12K Medium (Kaighn's Modification of Ham's F-12 Medium) contains 2 mM L-glutamine and 1500 mg/L sodium bicarbonate. It has applications for cell culture, cell growth, and viability.