Learn More
enQuire Bioreagents Calponin-1 Anti-Human Monoclonal [Clone SPM169], Size: 100ug
Supplier:  enQuire Bioreagents 2162MSM2P1
Encompass_Preferred
Product Type: Antibody. Application: IHC. Conjugate/Tag: Unconjugated. Clone: F13A1/1448. Clonality: Monoclonal. Host: Mouse. Isotype: IgG2b kappa. Positive Control: 293T Cells. Placenta. Cellular Staining: Cytoplasmic and Secreted. Buffer: 10mM PBS with 0.05% BSA and 0.05% Azide. Concentration: 200 ug/ml. Purity: Protein A/G Purified. Immunogen: Recombinant fragment of human Factor XIIIa protein (aa46-181). Storage: Short term: Store at 2-4C. Long term, aliquot and store at -20C. MW: 83kDa.Chromosome: 6p25.3-p24.3. Gene Symbol: F13A1. Gene ID: 2162. Ensemble: ENSG00000124491. Uniprot: NP_000120.
Catalog No. 50-188-4677