Learn More
Laser Vision Usa F19 Frame with P5E01 Laser Safety Filter
SDP
Supplier:  Laser Vision Usa F19.P5E01.5000
Encompass_Preferred
F19 Frame with P5E01 Laser Safety Filter, F19.P5E01.5000, Material: Polycarbonate, Color: Orange, VLT: 40%
Catalog No. NC2951600